Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethylcyclohexan-1-ol, 95%, Thermo Scientific™

Catalog No. BTBG00026EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTBG00026EA 10 g Amber Glass Bottle
BTBG00026DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTBG00026EA Supplier Thermo Scientific Chemicals Supplier No. BTBG00026EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 932-01-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00101954
InChI Key VUQOIZPFYIVUKD-UHFFFAOYSA-N
PubChem CID 136735
IUPAC Name 4,4-dimethylcyclohexan-1-ol
SMILES CC1(C)CCC(O)CC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 4,4-dimethylcyclohexanol, cyclohexanol, 4,4-dimethyl, 4,4-dimethyl-cyclohexanol, cyclohexanol,4,4-dimethyl, 4,4-dimethyl-1-cyclohexanol, 4,4-dimethyl cyclohexan-1-ol
Formula Weight 128.21
Percent Purity 95%
Chemical Name or Material 4, 4-Dimethylcyclohexan-1-ol
Hazard Category Warning
Hazard Statement GHS P Statement Causes serious eye irritation. Causes skin irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.