Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethyltriphenylamine 98.0+%, TCI America™
SDP

Catalog No. D23475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23475G 5 g
D23471G 1 g
D234725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D23475G Supplier TCI America Supplier No. D23475G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20440-95-3
Molecular Formula C20H19N
Molecular Weight (g/mol) 273.379
MDL Number MFCD00145004
InChI Key YWKKLBATUCJUHI-UHFFFAOYSA-N
Synonym Bis(4-methylphenyl)phenylamine, N,N-Di-p-tolylaniline, Phenyl(di-p-tolyl)amine
PubChem CID 88539
IUPAC Name 4-methyl-N-(4-methylphenyl)-N-phenylaniline
SMILES CC1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 5 g
Formula Weight 273.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethyltriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.