Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dinonyl-2,2'-bipyridyl 98.0+%, TCI America™
SDP

Catalog No. D39175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39175G 5 g
D39171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39175G Supplier TCI America Supplier No. D39175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 142646-58-0 | C28H44N2 | 408.674 g/mol

Chemical Identifiers

CAS 142646-58-0
Molecular Formula C28H44N2
Molecular Weight (g/mol) 408.674
MDL Number MFCD00800915
InChI Key VHJFWJXYEWHCGD-UHFFFAOYSA-N
Synonym 4,4'-dinonyl-2,2'-bipyridine, 4,4'-dinonyl-2,2'-bipyridyl, 4-nonyl-2-4-nonylpyridin-2-yl pyridine, 2,2'-bipyridine, 4,4'-dinonyl, 4,4'-bis nonyl-2,2'-bipyridine, dnbpy, acmc-1cctu, bpdn01, 4,4-dinonyl-2,2-bipyridine, 4,4-dinoyl-2,2-dipyridyl
PubChem CID 4285174
IUPAC Name 4-nonyl-2-(4-nonylpyridin-2-yl)pyridine
SMILES CCCCCCCCCC1=CC(=NC=C1)C2=NC=CC(=C2)CCCCCCCCC

Specifications

Melting Point 63°C
Color White
Quantity 5 g
Formula Weight 408.67
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dinonyl-2,2′-bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.