Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dinonyloxyazoxybenzene, TCI America™
SDP

Catalog No. D1518100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1518100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1518100MG Supplier TCI America Supplier No. D1518100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25729-13-9
Molecular Formula C30H46N2O3
Molecular Weight (g/mol) 482.709
MDL Number MFCD00059449
InChI Key KJMHEFPNHRGGGG-UHFFFAOYSA-N
PubChem CID 4575982
IUPAC Name (4-nonoxyphenyl)-(4-nonoxyphenyl)imino-oxidoazanium
SMILES CCCCCCCCCOC1=CC=C(C=C1)N=[N+](C2=CC=C(C=C2)OCCCCCCCCC)[O-]

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 482.71
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dinonyloxyazoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.