Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,4-diphenylbutylamine hydrochloride | 22101-90-2 | 99.0% | 261.79 g/mol | C16H20ClN | 5 MG
SDP

Catalog No. 5000222192
Encompass_Preferred

4,4-Diphenylbutylamine hydrochloride is the hydrochloride salt of 4,4-diphenylbutylamine and is used as a research reagent for studying serotonin (5-HT2A) and histamine (H1) receptor interactions. It is supplied as a characterized solid with reported purity and molecular properties suitable for biochemical assays.

  • White to off-white solid form, suitable for storage and handling.
  • Purity 99.0% supports reproducible biochemical results.
  • Reported Ki values indicate affinity for 5-HT2A (2589 nM) and H1 (1670 nM).
  • Molecular weight 261.79 g/mol and formula C16H20ClN for formulation calculations.
  • Available in small-mass formats and as DMSO solution for assay flexibility.

Catalog No. 50-002-22192 Supplier Medchemexpress LLC Supplier No. HY141422A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4,4-Diphenylbutylamine hydrochloride is the hydrochloride salt of 4,4-diphenylbutylamine and is used as a research reagent for studying serotonin (5-HT2A) and histamine (H1) receptor interactions. It is supplied as a characterized solid with reported purity and molecular properties suitable for biochemical assays.

  • White to off-white solid form, suitable for storage and handling.
  • Purity 99.0% supports reproducible biochemical results.
  • Reported Ki values indicate affinity for 5-HT2A (2589 nM) and H1 (1670 nM).
  • Molecular weight 261.79 g/mol and formula C16H20ClN for formulation calculations.
  • Available in small-mass formats and as DMSO solution for assay flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.