Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dipyridyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. D14835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D14835G Supplier TCI America Supplier No. D14835G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37968-97-1
Molecular Formula C10H8N2S
Molecular Weight (g/mol) 188.248
MDL Number MFCD00059784
InChI Key XGJOFCCBFCHEHK-UHFFFAOYSA-N
Synonym 4,4′C-Thiodipyridine
PubChem CID 604650
IUPAC Name 4-pyridin-4-ylsulfanylpyridine
SMILES C1=CN=CC=C1SC2=CC=NC=C2

Specifications

Melting Point 69°C
Quantity 5 g
Formula Weight 188.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dipyridyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.