Learn More
Medchemexpress LLC 4,4'-Disulfanediylbis(2-aminobutanoic acid) | 462-10-2 | 95.0% | 268.35 | 5 G
SDP
Catalog No. 5000366984
Encompass_Preferred
4,4'-Disulfanediylbis(2-aminobutanoic acid) is an endogenous metabolite provided by MedChemExpress for research use only, not for sale to patients. It is a white to off-white solid that requires specific storage conditions.
  • Endogenous metabolite.
  • For research use only.
  • White to off-white solid appearance.
  • Store at 4°C, protected from light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
Catalog No. 50-003-66984 Supplier Medchemexpress LLC Supplier No. HYW009390A5G
Add to Cart
Add to Cart
4,4'-Disulfanediylbis(2-aminobutanoic acid) is an endogenous metabolite provided by MedChemExpress for research use only, not for sale to patients. It is a white to off-white solid that requires specific storage conditions.
  • Endogenous metabolite.
  • For research use only.
  • White to off-white solid appearance.
  • Store at 4°C, protected from light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.