Learn More
Medchemexpress LLC 4-[4-[(E)-(2-oxo-1H-indol-3-ylidene)methyl]phenyl]piperazine-1-carbaldehyde | 186610-89-9 | 99.7% | 333.38 | C20H19N3O2 | 50 MG
SDP
Catalog No. 5000226700
Encompass_Preferred
SU4984 is a small-molecule, ATP-competitive protein tyrosine kinase inhibitor that primarily targets fibroblast growth factor receptor 1 (FGFR1) and is used for cellular and cancer research. The compound has formula C20H19N3O2 and a molecular weight of 333.38.
  • Targets FGFR1 with a reported IC50 of 10-20 μM.
  • Also inhibits platelet-derived growth factor receptor and insulin receptor.
  • Reported high purity by the supplier (99.7%).
  • Available in multiple pack sizes, including a 50 mg pack.
  • Recommended storage: solid at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month), protect from light.
Catalog No. 50-002-26700 Supplier Medchemexpress LLC Supplier No. HY11820350MG
SU4984 is a small-molecule, ATP-competitive protein tyrosine kinase inhibitor that primarily targets fibroblast growth factor receptor 1 (FGFR1) and is used for cellular and cancer research. The compound has formula C20H19N3O2 and a molecular weight of 333.38.
  • Targets FGFR1 with a reported IC50 of 10-20 μM.
  • Also inhibits platelet-derived growth factor receptor and insulin receptor.
  • Reported high purity by the supplier (99.7%).
  • Available in multiple pack sizes, including a 50 mg pack.
  • Recommended storage: solid at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month), protect from light.