Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Ethoxyphenoxycarbonyl)phenyl Ethyl Carbonate 98.0+%, TCI America™
SDP

Catalog No. E02571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E02571G Supplier TCI America Supplier No. E02571G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33926-25-9
Molecular Formula C18H18O6
Molecular Weight (g/mol) 330.336
MDL Number MFCD00059379
InChI Key NRRSSIQKLMBWJB-UHFFFAOYSA-N
Synonym Ethyl 4-(4-Ethoxyphenoxycarbonyl)phenyl Carbonate
PubChem CID 44630277
IUPAC Name (4-ethoxyphenyl) 4-ethoxycarbonyloxybenzoate
SMILES CCOC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)OC(=O)OCC

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 1 g
Formula Weight 330.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Ethoxyphenoxycarbonyl)phenyl Ethyl Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.