Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Ethoxyphenylazo)-m-phenylenediamine, mixture of mono and dihydrochloride, 98%

Catalog No. AC409680100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409680100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC409680100 Supplier Thermo Scientific Chemicals Supplier No. 409680100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 74367-87-6 | C14H16N4O·xHCl | 256.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74367-87-6
Molecular Formula C14H16N4O·xHCl
Molecular Weight (g/mol) 256.3
MDL Number MFCD00042023
Synonym ethoxazene hydrochloride, ethoxazene hcl, 4-4-ethoxyphenylazo-m-phenylenediamine monohydrochloride, etoxazene hydrochloride, ethoxy red, unii-5c90pjn6e5, ethoxazene hydrochloride usan, 4-4-ethoxyphenyl diazenyl benzene-1,3-diamine hydrochloride, 4-4-ethoxyphenylazo-1,3-phenylenediamine monohydrochloride, 1,3-benzenediamine, 4-4-ethoxyphenyl azo-, monohydrochloride

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 215°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Merck Index 12, 3796
Solubility Information Solubility in water: insoluble
Formula Weight 256.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Ethoxyphenylazo)-m-phenylenediamine, mixture of mono and dihydrochloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.