Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Ethylidenebisphenol 98.0+%, TCI America™
SDP

Catalog No. E043225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E043225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E043225G Supplier TCI America Supplier No. E043225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2081-08-5
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00191431
InChI Key HCNHNBLSNVSJTJ-UHFFFAOYSA-N
Synonym 1,1-Bis(4-hydroxyphenyl)ethane, 4,4′C-Ethylidenediphenol
PubChem CID 608116
ChEBI CHEBI:34029
IUPAC Name 4-[1-(4-hydroxyphenyl)ethyl]phenol
SMILES CC(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 25 g
Formula Weight 214.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Ethylidenebisphenol
EINECSNumber (4)-1550
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.