Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(4-fluorobenzoyl)-1-(4-phenylbutyl)piperidine oxalate | 144734-36-1 | 98.9% | 429.48 | C24H28FNO5 | 10 MG
SDP

Catalog No. 5000212764
Encompass_Preferred

4F 4PP oxalate is a research-grade selective 5-HT2A receptor antagonist with high affinity (Ki = 5.3 nM) and substantially lower affinity for 5-HT2C (Ki = 620 nM). Supplied as the oxalate salt, it is provided as a solid or as a DMSO solution for in vitro and in vivo studies; handle and store according to research-use guidance.

  • Selective 5-HT2A antagonist with high affinity.
  • Markedly lower affinity at 5-HT2C receptors.
  • High purity (98.86%).
  • Molecular weight 429.48; formula C24H28FNO5.
  • Soluble in DMSO; formulation guidance provided for in vivo use.
  • Available as solids and as a 10 mM DMSO solution in multiple sizes.
  • White to off-white solid; store sealed away from moisture.

Catalog No. 50-002-12764 Supplier Medchemexpress LLC Supplier No. HY10097010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4F 4PP oxalate is a research-grade selective 5-HT2A receptor antagonist with high affinity (Ki = 5.3 nM) and substantially lower affinity for 5-HT2C (Ki = 620 nM). Supplied as the oxalate salt, it is provided as a solid or as a DMSO solution for in vitro and in vivo studies; handle and store according to research-use guidance.

  • Selective 5-HT2A antagonist with high affinity.
  • Markedly lower affinity at 5-HT2C receptors.
  • High purity (98.86%).
  • Molecular weight 429.48; formula C24H28FNO5.
  • Soluble in DMSO; formulation guidance provided for in vivo use.
  • Available as solids and as a 10 mM DMSO solution in multiple sizes.
  • White to off-white solid; store sealed away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.