Learn More
Medchemexpress LLC 4-(4-fluoronaphthalen-1-yl)-6-propan-2-ylpyrimidin-2-amine | 199864-87-4 | MFCD20270616 | 100.0% | 281.33 g/mol | C17H16FN3 | 10 MG
SDP
Catalog No. 5000287428
Encompass_Preferred
RS-127445 is a selective, high-affinity 5-HT2B receptor antagonist used in preclinical research. It is orally bioavailable, shows a pKi of 9.5, and is supplied as a white to off-white solid suitable for both in vitro and in vivo studies. Intended for laboratory research use only.
  • Selective 5-HT2B receptor antagonist with nanomolar affinity.
  • Orally bioavailable and active in vivo models.
  • High chemical purity suitable for analytical and biological assays.
  • Available as a solid for convenient weighing and formulation.
  • Suitable for in vitro potency and selectivity profiling.
Catalog No. 50-002-87428 Supplier Medchemexpress LLC Supplier No. HY15419A10MG
RS-127445 is a selective, high-affinity 5-HT2B receptor antagonist used in preclinical research. It is orally bioavailable, shows a pKi of 9.5, and is supplied as a white to off-white solid suitable for both in vitro and in vivo studies. Intended for laboratory research use only.
  • Selective 5-HT2B receptor antagonist with nanomolar affinity.
  • Orally bioavailable and active in vivo models.
  • High chemical purity suitable for analytical and biological assays.
  • Available as a solid for convenient weighing and formulation.
  • Suitable for in vitro potency and selectivity profiling.