Learn More
Medchemexpress LLC 4-(4-fluorophenyl)-2-(4-nitrophenyl)-5-(4-pyridyl)-1H-imidazole | 152121-53-4 | MFCD12405019 | >98.0% | 360.34 g/mol | C20H13FN4O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10578R25MG
Encompass_Preferred
PD 169316 standard is an analytical reference material for a selective p38 MAP kinase inhibitor (CAS 152121-53-4), supplied as a high-purity solid for research and analytical applications such as compound identification, quantification, and method validation.
  • High purity suitable for analytical use.
  • Solid form for easy storage and handling.
  • Appropriate for assay calibration and method validation.
  • Compatible with chromatographic and mass spectrometry workflows.
  • Provided in small package sizes for trace-level analyses.
Catalog No. 50-002-40703