Learn More
Medchemexpress LLC 4-(4-fluorophenyl)-2-(4-nitrophenyl)-5-(4-pyridyl)-1H-imidazole | 152121-53-4 | MFCD12405019 | 98.6% | 360.34 g·mol⁻¹ | C20H13FN4O2 | 5 MG
SDP
Catalog No. 5000240234
Encompass_Preferred
PD 169316 is a potent, cell-permeable, and selective inhibitor of p38 mitogen-activated protein kinase (MAPK) with an IC50 of approximately 89 nM. Supplied as a high-purity research standard, it is used in biochemical and cell-based studies and has reported antiviral activity against Enterovirus 71.
  • Selective inhibition of p38 MAPK with ~89 nM IC50.
  • High purity suitable for analytical and biological assays.
  • Available as powder and as a DMSO solution for flexible use.
  • Recommended storage conditions extend shelf life for long-term studies.
  • Applicable to kinase signaling and antiviral research workflows.
Catalog No. 50-002-40234 Supplier Medchemexpress LLC Supplier No. HY10578R5MG
PD 169316 is a potent, cell-permeable, and selective inhibitor of p38 mitogen-activated protein kinase (MAPK) with an IC50 of approximately 89 nM. Supplied as a high-purity research standard, it is used in biochemical and cell-based studies and has reported antiviral activity against Enterovirus 71.
  • Selective inhibition of p38 MAPK with ~89 nM IC50.
  • High purity suitable for analytical and biological assays.
  • Available as powder and as a DMSO solution for flexible use.
  • Recommended storage conditions extend shelf life for long-term studies.
  • Applicable to kinase signaling and antiviral research workflows.