Learn More
Medchemexpress LLC 4-(4-Fluorophenyl)benzoic acid | 5731-10-2 | 99.0% | 216.21 | 1 ML
SDP
Catalog No. 5000345514
Encompass_Preferred
4-(4-Fluorophenyl)benzoic acid, also known as Compound 6, is a Bcl-2 binder with a KD value of 400 μM. It has potential for developing Bcl-2 selective anti-cancer agents.
  • Used for research purposes only.
  • Can be used to develop Bcl-2 selective anti-cancer agents.
Catalog No. 50-003-45514 Supplier Medchemexpress LLC Supplier No. HY10961750MG
4-(4-Fluorophenyl)benzoic acid, also known as Compound 6, is a Bcl-2 binder with a KD value of 400 μM. It has potential for developing Bcl-2 selective anti-cancer agents.
  • Used for research purposes only.
  • Can be used to develop Bcl-2 selective anti-cancer agents.