Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Formylphenyl)morpholine 98.0+%, TCI America™
SDP

Catalog No. M16255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16255G 5 g
M16251G 1 g
M162525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M16255G Supplier TCI America Supplier No. M16255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1204-86-0
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00735826
InChI Key FOAQOAXQMISINY-UHFFFAOYSA-N
Synonym 4-morpholinobenzaldehyde, 4-morpholin-4-yl-benzaldehyde, 4-4-morpholinyl benzaldehyde, 4-4-formylphenyl morpholine, 4-morpholin-4-yl benzaldehyde, 4-morpholinobenzenecarbaldehyde, benzaldehyde, 4-4-morpholinyl, akos bc-2976, acmc-1bqjc, 4-morpholinylbenzaldehyde
PubChem CID 291349
IUPAC Name 4-(morpholin-4-yl)benzaldehyde
SMILES O=CC1=CC=C(C=C1)N1CCOCC1

Specifications

Melting Point 63°C
Quantity 5 g
Formula Weight 191.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Formylphenyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.