Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde

Catalog No. 95594100MGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
95-594-100MGF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 95-594-100MGF Supplier Sigma Aldrich Supplier No. 95594100MGF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-formylphenyl sulfofluoridate is an aryl fluorosulfate
  • It can be synthesized from the reaction between phenol and sulfuryl fluoride in the presence of triethylamine
  • It undergoes sonogashira coupling reaction with phenyl acetylene.the following aryl fluorosulfate can be utilized as a cross-coupling partner in suzuki-miyaura reactions
  • The standard reaction conditions for these palladium-catalyzed c-c bond formations are ligand-free and can be performed in water, at room temperature and are not air sensitive.palladium-catalyzed, ligand-free suzuki reaction in water using aryl fluorosulfates

Specifications

CAS 31906-04-4
Density 0.995 g/mL (at 20°C)
Molecular Formula C13H22O2
Linear Formula C13H22O2
MDL Number MFCD00019423
Quantity 1 g
Molecular Weight (g/mol) 210.31
Percent Purity ≥97.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.