Learn More
Medchemexpress LLC 4-(4-hydroxyphenyl)-2-butanone | 5471-51-2 | 2394 | 164.20 | 100 MG
SDP
Catalog No. 5000416109
Encompass_Preferred
Raspberry ketone (Standard) is the analytical standard of Raspberry ketone, a major aromatic compound found in red raspberry. It is commonly used as a fragrance in cosmetics and a flavoring agent in food, and also exhibits PPAR-α agonistic activity.
  • Intended for research and analytical applications.
  • Reference standard for qualitative, quantitative, and methodological research experiments.
  • Suitable for techniques such as HPLC, GC, and MS.
Catalog No. 50-004-16109 Supplier Medchemexpress LLC Supplier No. HYN1426R100MG
Raspberry ketone (Standard) is the analytical standard of Raspberry ketone, a major aromatic compound found in red raspberry. It is commonly used as a fragrance in cosmetics and a flavoring agent in food, and also exhibits PPAR-α agonistic activity.
  • Intended for research and analytical applications.
  • Reference standard for qualitative, quantitative, and methodological research experiments.
  • Suitable for techniques such as HPLC, GC, and MS.