Learn More
Medchemexpress LLC 4-(4-hydroxyphenyl)-2-butanone | 5471-51-2 | MFCD00002394 | 100.0% | 164.20 g/mol | C10H12O2 | 10 MG
SDP
Catalog No. 5000296130
Encompass_Preferred
Raspberry ketone (4-(4-hydroxyphenyl)-2-butanone) is provided as an analytical reference standard in a small, accurately weighed solid format for laboratory research. It is intended for method development, calibration, and quality control in analytical workflows such as chromatography and mass spectrometry.
  • Analytical standard suitable for method development and quality control.
  • High purity for consistent, reliable analytical results.
  • Solid, white to off-white appearance; easy to handle and weigh.
  • Compatible with chromatographic and spectrometric assays for reference and calibration.
Catalog No. 50-002-96130 Supplier Medchemexpress LLC Supplier No. HYN1426R10MG
Raspberry ketone (4-(4-hydroxyphenyl)-2-butanone) is provided as an analytical reference standard in a small, accurately weighed solid format for laboratory research. It is intended for method development, calibration, and quality control in analytical workflows such as chromatography and mass spectrometry.
  • Analytical standard suitable for method development and quality control.
  • High purity for consistent, reliable analytical results.
  • Solid, white to off-white appearance; easy to handle and weigh.
  • Compatible with chromatographic and spectrometric assays for reference and calibration.