Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Methyl-1,3-dioxolan-2-yl)-1,2-methylenedioxybenzene 98.0+%, TCI America™
SDP

Catalog No. M248325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M248325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M248325G Supplier TCI America Supplier No. M248325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61683-99-6
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD13194964
InChI Key HXNSADTWQWLFCB-UHFFFAOYSA-N
Synonym 5-(4-Methyl-1,3-dioxolan-2-yl)-1,3-benzodioxole, Piperonal Propylene Glycol Acetal
PubChem CID 101144
IUPAC Name 5-(4-methyl-1,3-dioxolan-2-yl)-1,3-benzodioxole
SMILES CC1COC(O1)C2=CC3=C(C=C2)OCO3

Specifications

Color Colorless
Quantity 25 g
Formula Weight 208.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(4-Methyl-1,3-dioxolan-2-yl)-1,2-methylenedioxybenzene
EINECSNumber (5)-0512
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.