Learn More
Medchemexpress LLC β-[4-([4-methyl-2-(4-(trifluoromethyl)phenyl)-5-thiazolyl]methoxy)phenyl]-3-isoxazolepropanoic acid | 1011531-27-3 | 99.0% | 488.48 | C24H19F3N2O4S | 10MG
SDP
Supplier:  Medchemexpress LLC HY1258510MG
Encompass_Preferred
AM-4668 is a research-grade small-molecule agonist of GPR40 (FFAR1) used in preclinical studies relevant to type 2 diabetes. It activates human GPR40 with low-nanomolar potency in cell-based functional assays and is supplied as a solid for in vitro pharmacology and biochemical studies.
  • Low-nanomolar potency at GPR40 (EC50 3.6 nM in A9 cells, 36 nM in CHO cells).
  • Suitable for in vitro receptor activation and pharmacology studies.
  • High purity solid (99.0%).
  • Molecular weight 488.48 and formula C24H19F3N2O4S.
  • Available in milligram quantities for laboratory use.
Catalog No. 50-002-38246