Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4'-Methylene-bis(2-chloroaniline)

Catalog No. 117323250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-732-3250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-732-3250G Supplier Sigma Aldrich Supplier No. 117323250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4'-Methylene-bis(2-chloroaniline)

Specifications

CAS 101-14-4
Melting Point 102°C to 107°C
Boiling Point 202°C to 214°C/0.3 mmHg (literature)
Molecular Formula C13H12Cl2N2
Linear Formula CH2[C6H3(Cl)NH2]2
Quantity 250 g
MDL Number MFCD00047829
Synonym 2,2 -Dichloro-4,4 -methylenedianiline; 4,4 -Diamino-3,3 -dichlorodiphenylmethane
Molecular Weight (g/mol) 267.15
Percent Purity 85%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.