Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4'-Methylenebis(2-methylcyclohexylamine), mixture of isomers

Catalog No. 369500250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-950-0250ML 250 mL
36-950-01L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-950-0250ML Supplier Sigma Aldrich Supplier No. 369500250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4,4'-methylenebis(2-methylcyclohexylamine) is an alicyclic diamine.4,4α-methylenebis(2-methylcyclohexylamine) (mmca) may be used:
  • In the synthesis of fully aliphatic polyimides
  • In the synthesis of fully aliphatic polyimide-siloxanes (apisio)
  • To prepare epoxy resin with trimethylolpropane triglycidyl ether, used for the characterization of the structural organization in dragline silk by transmission x-ray microscopy (stxm)

Specifications

CAS 6864-37-5
Density 0.94 g/mL (at 25°C (literature))
Boiling Point 93°C to 100°C (lit.)
Molecular Formula C15H30N2
Refractive Index n20/D 1.499 (literature)
Linear Formula CH2[C6H9(CH3)NH2]2
Quantity 250 mL
MDL Number MFCD00075535
Molecular Weight (g/mol) 238.41
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.