Learn More
Medchemexpress LLC 4-[(4-methylphenyl)thio]thieno[2,3-c]pyridine-2-carboxamide | 251992-66-2 | MFCD09038566 | ≥98.0% | 300.4 g/mol | C15H12N2OS2 | 50 MG
SDP
Catalog No. 5000215626
Encompass_Preferred
A-205804 is a small-molecule research inhibitor that selectively reduces E-selectin and ICAM-1 expression and is used in preclinical studies of inflammatory and immune-related pathways. It demonstrates low-nanomolar potency in cellular assays and is typically supplied as a solid for formulation and dosing in laboratory research.
  • Potent inhibitor of E-selectin and ICAM-1 (IC50 ≈ 20 nM and 25 nM)
  • Orally bioavailable small molecule
  • Suitable for in vitro and in vivo preclinical research
  • Molecular formula C15H12N2OS2 and molecular weight ≈ 300.4 g/mol
  • Supplied as solid material for formulation and dosing
Catalog No. 50-002-15626 Supplier Medchemexpress LLC Supplier No. HY10022650MG
A-205804 is a small-molecule research inhibitor that selectively reduces E-selectin and ICAM-1 expression and is used in preclinical studies of inflammatory and immune-related pathways. It demonstrates low-nanomolar potency in cellular assays and is typically supplied as a solid for formulation and dosing in laboratory research.
  • Potent inhibitor of E-selectin and ICAM-1 (IC50 ≈ 20 nM and 25 nM)
  • Orally bioavailable small molecule
  • Suitable for in vitro and in vivo preclinical research
  • Molecular formula C15H12N2OS2 and molecular weight ≈ 300.4 g/mol
  • Supplied as solid material for formulation and dosing