Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Methylpiperazino)benzoic acid, 97%, Thermo Scientific™

Catalog No. CC18701CB
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC18701CB 250 mg Amber Glass Bottle
CC18701DA 1 g Amber Glass Bottle
CC18701EB 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC18701CB Supplier Thermo Scientific Chemicals Supplier No. CC18701CB
Add to Cart

Chemical Identifiers

CAS 86620-62-4
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD02682063
InChI Key UCFZVQHKTRSZMM-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl benzoic acid, 4-4-methylpiperazino benzoic acid, 4-4-methyl-piperazin-1-yl-benzoic acid, 4-4-methylpiperazinyl benzoic acid, 4-4-methyl-piperazino benzoic acid, benzoic acid, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzoic acid, pubchem10481, ksc448o3p
PubChem CID 736532
IUPAC Name 4-(4-methylpiperazin-1-yl)benzoic acid
SMILES CN1CCN(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 4-(4-Methylpiperazino)benzoic acid
Hazard Category Warning
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.