Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,4'-octanamidostilbene-2,2'-disulfonate | 5970-15-0 | >98.0% | 25 MG
SDP

Catalog No. 5000231467
Encompass_Preferred

OADS (4,4'-octanamidostilbene-2,2'-disulfonate) is a small-molecule inhibitor of the ClC-ec1 Cl-/H+ antiporter used as a biochemical and structural research reagent.

  • Inhibits ClC-ec1 with low micromolar affinity.
  • Designed disulfonated stilbene with octanamide side chains.
  • CAS number 5970-15-0.
  • Purity greater than 98%.
  • Molecular weight 666.76 g/mol; formula C30H40N2Na2O8S2.
  • For research use only; not for human or veterinary use.
  • Typical package size 25 MG.

Catalog No. 50-002-31467 Supplier Medchemexpress LLC Supplier No. HY12273225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OADS (4,4'-octanamidostilbene-2,2'-disulfonate) is a small-molecule inhibitor of the ClC-ec1 Cl-/H+ antiporter used as a biochemical and structural research reagent.

  • Inhibits ClC-ec1 with low micromolar affinity.
  • Designed disulfonated stilbene with octanamide side chains.
  • CAS number 5970-15-0.
  • Purity greater than 98%.
  • Molecular weight 666.76 g/mol; formula C30H40N2Na2O8S2.
  • For research use only; not for human or veterinary use.
  • Typical package size 25 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.