Learn More
Medchemexpress LLC 4-((4-oxo-2-thioxo-3-(3-(trifluoromethyl)benzyl)thiazolidin-5-ylidene)methyl)benzoic acid | 1073612-91-5 | MFCD31619349 | 99.6% | 423.43 g/mol | C19H12F3NO3S2 | 25 MG
SDP
Catalog No. 5000245045
Encompass_Preferred
CY-09 is a selective, direct inhibitor of the NLRP3 inflammasome that binds the ATP-binding motif of the NLRP3 NACHT domain and inhibits NLRP3 ATPase activity, suppressing inflammasome assembly and activation. Supplied as a high-purity solid for research use.
  • Selective NLRP3 inhibitor targeting the ATP-binding motif
  • Inhibits NLRP3 ATPase activity to block inflammasome activation
  • High purity solid form for research applications
  • Light yellow to yellow appearance
  • Stable when stored at recommended low temperatures
  • Available in small research sizes, including 25 mg
Catalog No. 50-002-45045 Supplier Medchemexpress LLC Supplier No. HY10366625MG
CY-09 is a selective, direct inhibitor of the NLRP3 inflammasome that binds the ATP-binding motif of the NLRP3 NACHT domain and inhibits NLRP3 ATPase activity, suppressing inflammasome assembly and activation. Supplied as a high-purity solid for research use.
  • Selective NLRP3 inhibitor targeting the ATP-binding motif
  • Inhibits NLRP3 ATPase activity to block inflammasome activation
  • High purity solid form for research applications
  • Light yellow to yellow appearance
  • Stable when stored at recommended low temperatures
  • Available in small research sizes, including 25 mg