Learn More
Medchemexpress LLC 4-[[4-oxo-2-thioxo-3-[[3-(trifluoromethyl)phenyl]methyl]-5-thiazolidinylidene]methyl]benzoic acid | 1073612-91-5 | MFCD31619349 | 99.6% | 423.43 g/mol | C19H12F3NO3S2 | 1 MG
SDP
Catalog No. 5000246902
Encompass_Preferred
Selective, direct small-molecule inhibitor of the NLRP3 inflammasome that binds the ATP-binding motif of the NLRP3 NACHT domain and inhibits NLRP3 ATPase activity. Supplied as a light-yellow solid and provided in milligram-scale solids or pre-prepared DMSO solution formats for research applications.
  • Selective NLRP3 inhibitor that blocks ATPase activity.
  • Direct binding to the ATP-binding motif of the NLRP3 NACHT domain.
  • High stated purity (99.6%).
  • Light yellow solid; soluble in DMSO for solution preparation.
  • Available in milligram-scale solids and 10 mM DMSO solutions.
  • Intended for in vitro and in vivo research; not for human use.
Catalog No. 50-002-46902 Supplier Medchemexpress LLC Supplier No. HY1036661MG
Selective, direct small-molecule inhibitor of the NLRP3 inflammasome that binds the ATP-binding motif of the NLRP3 NACHT domain and inhibits NLRP3 ATPase activity. Supplied as a light-yellow solid and provided in milligram-scale solids or pre-prepared DMSO solution formats for research applications.
  • Selective NLRP3 inhibitor that blocks ATPase activity.
  • Direct binding to the ATP-binding motif of the NLRP3 NACHT domain.
  • High stated purity (99.6%).
  • Light yellow solid; soluble in DMSO for solution preparation.
  • Available in milligram-scale solids and 10 mM DMSO solutions.
  • Intended for in vitro and in vivo research; not for human use.