Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Oxybis(benzenesulfonyl Chloride) 97.0+%, TCI America™
SDP

Catalog No. O029825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O029825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O029825G Supplier TCI America Supplier No. O029825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121-63-1
Molecular Formula C12H8Cl2O5S2
Molecular Weight (g/mol) 367.21
MDL Number MFCD00024884
InChI Key HJKXLQIPODSWMB-UHFFFAOYSA-N
Synonym 4,4'-bis chlorosulfonyl diphenyl ether, bis 4-chlorosulfonylphenyl ether, 4,4'-oxybis benzenesulfonyl chloride, 4,4'-oxydibenzenesulfonyl chloride, oxybis benzenesulfonyl chloride, 4,4'-oxybenzenesulfonylchloride, benzenesulfonyl chloride, 4,4'-oxybis, oxybis 4-benzenesulfonyl chloride, p,p'-oxybis benzenesulfonyl chloride, phenoxybenzene-4,4'-disulfonyl chloride
PubChem CID 8484
IUPAC Name 4-[4-(chlorosulfonyl)phenoxy]benzene-1-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)C=C1

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 367.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Oxybis(benzenesulfonyl Chloride)
EINECSNumber (3)-0847&(3)-3173
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.