Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Oxybis(benzenesulfonyl hydrazide), 95%, Thermo Scientific™

Catalog No. AAH6123936
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6123936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6123936 Supplier Thermo Scientific Chemicals Supplier No. H6123936

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4,4′-Oxybis(benzenesulfonyl hydrazide) is an organic hydrazide used as a blowing agent for sponge rubber and expanded plastics. Used as an additive to polymers to make sponge rubber cosmetic puffs, electrostatographic toners, buoyant seats, insulation and packing materials, adhesives, sealants, and leather substitutes.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-51-3
Molecular Formula C12H14N4O5S2
Molecular Weight (g/mol) 358.39
MDL Number MFCD00007587
InChI Key NBOCQTNZUPTTEI-UHFFFAOYSA-N
Synonym 4,4'-oxydibenzenesulfonohydrazide, 4,4'-oxybis benzenesulfonyl hydrazide, celogen, serogen, obsh, genitron ob, zhenitron ov, celogen ot, 4,4'-oxybis benzenesulfonhydrazide, cellmic s
PubChem CID 6649
IUPAC Name 4-[4-(hydrazinesulfonyl)phenoxy]benzenesulfonohydrazide
SMILES NNS(=O)(=O)C1=CC=C(OC2=CC=C(C=C2)S(=O)(=O)NN)C=C1

Specifications

Color White
Quantity 500 g
UN Number UN3226
Formula Weight 358.39
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4,4'-Oxybis(benzenesulfonyl hydrazide)
Hazard Category Danger
Hazard Statement GHS H Statement
H228-H242
Flammable solid.
Heating may cause a fire.
Precautionary Statement GHS P Statement
P210-P220-P280-P370+P378a-P411a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store at temperatures not exceeding 392°F.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.1; Packaging Group: II; Proper Shipping Name: SELF-REACTIVE SOLID TYPE D
RTECSNumber DB7321000
TSCA TSCA
storageNote1 -30° to -10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.