Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Oxybis(benzoyl Chloride) 98.0+%, TCI America™
SDP

Catalog No. O039925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O039925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O039925G Supplier TCI America Supplier No. O039925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7158-32-9
Molecular Formula C14H8Cl2O3
Molecular Weight (g/mol) 295.115
MDL Number MFCD00496619
InChI Key OSUWBBMPVXVSOA-UHFFFAOYSA-N
Synonym 4,4′C-Oxydibenzoyl Chloride, 4,4′C-Bis(chlorocarbonyl)diphenyl Ether, Bis(4-chlorocarbonylphenyl) Ether
PubChem CID 23546
IUPAC Name 4-(4-carbonochloridoylphenoxy)benzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)OC2=CC=C(C=C2)C(=O)Cl

Specifications

Melting Point 90°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 295.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Oxybis(benzoyl Chloride)
EINECSNumber (3)-4482
RTECSNumber DM6664000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.