Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,4'-((Pentane-1,5-diyl-1,1,5,5-d4)bis(oxy))dibenzimidamide dihydrochloride | 1276197-32-0 | 98.0% | 1 MG
SDP

Catalog No. 5000408847
Encompass_Preferred

Pentamidine-d4 dihydrochloride is the deuterium labeled Pentamidine dihydrochloride. It is an antimicrobial agent that interferes with DNA biosynthetics. This product appears as a white to off-white solid with a purity of 98.0% and an isotopic enrichment of 99.2%.

  • Deuterium labeled compound
  • Antimicrobial agent
  • Interferes with DNA biosynthetics
  • White to off-white solid
  • Purity 98.0%
  • Isotopic enrichment 99.2%
  • Recommended storage at 4°C, sealed, away from moisture
  • Recommended storage in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-08847 Supplier Medchemexpress LLC Supplier No. HYB0537AS1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Pentamidine-d4 dihydrochloride is the deuterium labeled Pentamidine dihydrochloride. It is an antimicrobial agent that interferes with DNA biosynthetics. This product appears as a white to off-white solid with a purity of 98.0% and an isotopic enrichment of 99.2%.

  • Deuterium labeled compound
  • Antimicrobial agent
  • Interferes with DNA biosynthetics
  • White to off-white solid
  • Purity 98.0%
  • Isotopic enrichment 99.2%
  • Recommended storage at 4°C, sealed, away from moisture
  • Recommended storage in solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.