Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,4'-((Pentane-1,5-diyl-1,1,5,5-d4)bis(oxy))dibenzimidamide dihydrochloride | 1276197-32-0 | 98.0% | 5 MG
SDP

Catalog No. 5000407621
Encompass_Preferred

This product is Pentamidine-d4 dihydrochloride, with a purity of 98.0%. It appears as a white to off-white solid. It has a molecular formula of C19H22D4Cl2N4O2 and a molecular weight of 417.37. It is intended for research use only.

  • Appearance: White to off-white solid
  • Purity: 98.0%
  • Isotopic enrichment: 99.2%
  • Molecular formula: C19H22D4Cl2N4O2
  • Molecular weight: 417.37
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Catalog No. 50-004-07621 Supplier Medchemexpress LLC Supplier No. HYB0537AS5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This product is Pentamidine-d4 dihydrochloride, with a purity of 98.0%. It appears as a white to off-white solid. It has a molecular formula of C19H22D4Cl2N4O2 and a molecular weight of 417.37. It is intended for research use only.

  • Appearance: White to off-white solid
  • Purity: 98.0%
  • Isotopic enrichment: 99.2%
  • Molecular formula: C19H22D4Cl2N4O2
  • Molecular weight: 417.37
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.