Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(4-(Prop-2-Yn-1-Yloxy)Benzoyl)Benzoic Acid

Catalog No. 900602100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-060-2100MG 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-060-2100MG Supplier Sigma Aldrich Supplier No. 900602100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-(4-(Prop-2-yn-1-yloxy)benzoyl)benzoic acid is used for chemical probe synthesis, this trifunctional building block contains a light-activated benzophenone, alkyne tag, and carboxylic acid synthetic handle. When appended to a ligand or pharmacophore through its acid linker, this building block allows for UV light-induced covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.Technology Spotlight: Trifuctional Probe Building BlocksA library approach to rapidly discover photoaffinity probes of the mRNA decapping scavenger enzyme DcpSSynthesis and Evaluation of New Thiodigalactoside-Based Chemical Probes to Label Galectin-3Discovery of tumor-specific irreversible inhibitors of stearoyl CoA desaturase

Specifications

CAS 2140866-80-2
Molecular Formula C17H12O4
Linear Formula C17H12O4
Quantity 10 g
Synonym 2,4,6-Trimercaptotriazine-functionalized silica gel; SiliaMetS(R) Dimercaptotriazine (DMT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.