Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Propylphenyl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. P103710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P103710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P103710G Supplier TCI America Supplier No. P103710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88038-94-2
Molecular Formula C16H16O2
Molecular Weight (g/mol) 240.302
MDL Number MFCD00017335
InChI Key HCPBURTZSXRGBN-UHFFFAOYSA-N
Synonym 4-4-propylphenyl benzoic acid, 4-4-n-propylphenyl benzoic acid, 4'-propyl-1,1'-biphenyl-4-carboxylic acid, 4-propylbiphenyl-4'-carboxylic acid, 4-propyl-4'-biphenylcarboxylic acid, 4'-propylbiphenyl-4-carboxylic acid, 4-n-propylbiphenyl-4'-carboxylic acid, 4-p-n-propylphenyl benzoic acid, 1,1'-biphenyl-4-carboxylic acid, 4'-propyl, 4'-n-propylbiphenyl-4-carboxylic acid
PubChem CID 522891
IUPAC Name 4-(4-propylphenyl)benzoic acid
SMILES CCCC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Color White-Yellow
Quantity 10 g
Formula Weight 240.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Propylphenyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.