Learn More
Medchemexpress LLC 4,4'-sulfonylbis(2-methylphenol) | 16346-97-7 | MFCD00130281 | 97.0% | 278.32 g/mol | C14H14O4S | 5 MG
SDP
Catalog No. 5000283062
Encompass_Preferred
This bisphenol-derived small-molecule inhibits DNA topoisomerase II alpha and is intended for research use. It demonstrates topo II inhibitory activity and in vitro cell growth inhibition, making it suitable for biochemical and anticancer studies.
  • Small-molecule inhibitor of DNA topoisomerase II alpha.
  • Bisphenol-derived structure; formula C14H14O4S.
  • Documented in vitro topo II inhibitory activity and cell growth IC50 values.
  • Supplied in small quantities for research use (5 mg pack).
  • Intended for biochemical and anticancer research applications.
Catalog No. 50-002-83062 Supplier Medchemexpress LLC Supplier No. HYW1780115MG
This bisphenol-derived small-molecule inhibits DNA topoisomerase II alpha and is intended for research use. It demonstrates topo II inhibitory activity and in vitro cell growth inhibition, making it suitable for biochemical and anticancer studies.
  • Small-molecule inhibitor of DNA topoisomerase II alpha.
  • Bisphenol-derived structure; formula C14H14O4S.
  • Documented in vitro topo II inhibitory activity and cell growth IC50 values.
  • Supplied in small quantities for research use (5 mg pack).
  • Intended for biochemical and anticancer research applications.