Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-tert-Butylphenyl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B19861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19861G 1 g
B19865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19861G Supplier TCI America Supplier No. B19861G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5748-42-5
Molecular Formula C17H18O2
Molecular Weight (g/mol) 254.329
MDL Number MFCD01631300
InChI Key HZIOPONJCVOCFE-UHFFFAOYSA-N
PubChem CID 541479
IUPAC Name 4-(4-tert-butylphenyl)benzoic acid
SMILES CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Synonym 4-tert-Butylbiphenyl-4′C-carboxylic Acid
Formula Weight 254.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-tert-Butylphenyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.