Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[4-(Trifluoromethoxy)phenoxy]piperidine 97.0+%, TCI America™
SDP

Catalog No. T33491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33491G 1 g
T33495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33491G Supplier TCI America Supplier No. T33491G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 287952-67-4
Molecular Formula C12H14F3NO2
Molecular Weight (g/mol) 261.24
MDL Number MFCD06656166
InChI Key RPQOTFPZKNHYFB-UHFFFAOYSA-N
Synonym 4-4-trifluoromethoxy phenoxy piperidine, 4-4-trifluoromethoxyphenoxy piperidine, piperidine, 4-4-trifluoromethoxy phenoxy, 4-4-trifluoromethoxy-phenoxy-piperidine, acmc-1cpm9, 4-4-trifluoromethoxy-phenoxy piperidine, 4-4-trifluoromethoxy phenoxy-piperidine
PubChem CID 16115439
IUPAC Name 4-[4-(trifluoromethoxy)phenoxy]piperidine
SMILES FC(F)(F)OC1=CC=C(OC2CCNCC2)C=C1

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 1 g
Formula Weight 261.24
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[4-(Trifluoromethoxy)phenoxy]piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.