Learn More
Medchemexpress LLC 4-(5-(4,7-dimethylbenzofuran-2-yl)-1,2,4-oxadiazol-3-yl)benzoic acid | 1952276-71-9 | 98.9% | 334.33 g·mol⁻1 | C19H14N2O4 | 5 MG
SDP
Catalog No. 5000263505
Encompass_Preferred
KCL-286 (C286) is a potent, selective retinoic acid receptor beta-2 (RARβ2) agonist reported to be orally active and brain-penetrant (reported EC50 = 1.9 nM). It is intended for research use in in vitro and in vivo studies of retinoid signaling and neurological pharmacology. Characterization includes molecular formula C19H14N2O4, molecular weight 334.33 g·mol⁻1, and CAS 1952276-71-9.
  • Potent RARβ2 agonist (EC50 = 1.9 nM).
  • Orally active and brain-penetrant in reported studies.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Characterized by molecular formula C19H14N2O4 and molecular weight 334.33 g·mol⁻1.
  • Supplied as a high-purity research reagent (≈98.9% reported purity).
Catalog No. 50-002-63505 Supplier Medchemexpress LLC Supplier No. HY1115735MG
KCL-286 (C286) is a potent, selective retinoic acid receptor beta-2 (RARβ2) agonist reported to be orally active and brain-penetrant (reported EC50 = 1.9 nM). It is intended for research use in in vitro and in vivo studies of retinoid signaling and neurological pharmacology. Characterization includes molecular formula C19H14N2O4, molecular weight 334.33 g·mol⁻1, and CAS 1952276-71-9.
  • Potent RARβ2 agonist (EC50 = 1.9 nM).
  • Orally active and brain-penetrant in reported studies.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Characterized by molecular formula C19H14N2O4 and molecular weight 334.33 g·mol⁻1.
  • Supplied as a high-purity research reagent (≈98.9% reported purity).