Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[5-(4-butylcyclohexyl)-1,2,4-oxadiazol-3-yl]pyridine | 388575-52-8 | MFCD01829706 | 100.0% | 285.38 | C17H23N3O | 25 MG
SDP

Catalog No. 5000242717
Encompass_Preferred

PSN 375963 is a small-molecule agonist of the G protein-coupled receptor GPR119 used as a research tool to study GPR119-mediated signaling and metabolic effects. It is supplied as a characterized solid for in vitro and preclinical investigations.

  • Potent GPR119 agonist with human EC50 8.4 μM and mouse EC50 7.9 μM.
  • Chemical formula C17H23N3O; molecular weight 285.38 g/mol.
  • Solid, white to off-white appearance, suitable for assay use.
  • High purity (≈99.96%) by HPLC.
  • Recommended storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C.

Catalog No. 50-002-42717 Supplier Medchemexpress LLC Supplier No. HY10825825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSN 375963 is a small-molecule agonist of the G protein-coupled receptor GPR119 used as a research tool to study GPR119-mediated signaling and metabolic effects. It is supplied as a characterized solid for in vitro and preclinical investigations.

  • Potent GPR119 agonist with human EC50 8.4 μM and mouse EC50 7.9 μM.
  • Chemical formula C17H23N3O; molecular weight 285.38 g/mol.
  • Solid, white to off-white appearance, suitable for assay use.
  • High purity (≈99.96%) by HPLC.
  • Recommended storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.