Learn More
Medchemexpress LLC 4-[5-(4-butylcyclohexyl)-1,2,4-oxadiazol-3-yl]pyridine | 388575-52-8 | MFCD01829706 | 100.0% | 285.38 g·mol⁻¹ | C17H23N3O | 100 MG
SDP
Catalog No. 5000242681
Encompass_Preferred
PSN 375963 is a small-molecule agonist of the G protein-coupled receptor GPR119 used as a research tool for in vitro pharmacology. It exhibits micromolar potency and is provided with supporting analytical documentation for use in biochemical and cell-based assays.
  • Shows micromolar agonist activity at GPR119 (EC50s 8.4 μM human, 7.9 μM mouse).
  • Supplied as a characterized research-grade compound with COA and SDS.
  • Available as solid powder and DMSO solutions for assay flexibility.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular formula C17H23N3O and molecular weight 285.38 g·mol⁻¹.
Catalog No. 50-002-42681 Supplier Medchemexpress LLC Supplier No. HY108258100MG
PSN 375963 is a small-molecule agonist of the G protein-coupled receptor GPR119 used as a research tool for in vitro pharmacology. It exhibits micromolar potency and is provided with supporting analytical documentation for use in biochemical and cell-based assays.
  • Shows micromolar agonist activity at GPR119 (EC50s 8.4 μM human, 7.9 μM mouse).
  • Supplied as a characterized research-grade compound with COA and SDS.
  • Available as solid powder and DMSO solutions for assay flexibility.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular formula C17H23N3O and molecular weight 285.38 g·mol⁻¹.