Learn More
Medchemexpress LLC 4-[5-(4-butylcyclohexyl)-1,2,4-oxadiazol-3-yl]pyridine | 388575-52-8 | MFCD01829706 | 100.0% | 285.38 g·mol⁻¹ | C17H23N3O | 5 MG
SDP
Catalog No. 5000242673
Encompass_Preferred
PSN 375963 is a small-molecule agonist of G protein-coupled receptor 119 (GPR119) used as a research tool to probe GPR119 signaling and metabolic pathways. It is reported with low micromolar activity and is provided as a solid suitable for biochemical and cellular assays.
  • Potent GPR119 agonist with EC50 ≈ 8.4 μM (human) and 7.9 μM (mouse).
  • Small molecule with formula C17H23N3O and molecular weight 285.38 g·mol⁻¹.
  • Supplied as a solid suitable for biochemical and cellular assays.
  • High reported purity (99.96%).
  • Used to study GPR119-mediated metabolic signaling and receptor pharmacology.
Catalog No. 50-002-42673 Supplier Medchemexpress LLC Supplier No. HY1082585MG
PSN 375963 is a small-molecule agonist of G protein-coupled receptor 119 (GPR119) used as a research tool to probe GPR119 signaling and metabolic pathways. It is reported with low micromolar activity and is provided as a solid suitable for biochemical and cellular assays.
  • Potent GPR119 agonist with EC50 ≈ 8.4 μM (human) and 7.9 μM (mouse).
  • Small molecule with formula C17H23N3O and molecular weight 285.38 g·mol⁻¹.
  • Supplied as a solid suitable for biochemical and cellular assays.
  • High reported purity (99.96%).
  • Used to study GPR119-mediated metabolic signaling and receptor pharmacology.