Learn More
Medchemexpress LLC 4-(5-(4-chlorophenyl)-4-methyl-2-propionylthiophen-3-yl)benzenesulfonamide | 1394371-75-5 | MFCD32671353 | 98.4% | 419.94 | C20H18ClNO3S2 | 5 MG
SDP
Catalog No. 5000262638
Encompass_Preferred
nAChR agonist 1 is a research compound that functions as a potent positive allosteric modulator of the α7 nicotinic acetylcholine receptor. It is brain-permeable and orally efficacious in preclinical assays, with reported activity in FLIPR Ca2+ mobilization studies. Supplied as an off-white to yellow solid for use in in vitro and in vivo research.
  • Potent positive allosteric modulator of α7 nicotinic acetylcholine receptor.
  • Brain-permeable and orally efficacious in preclinical models.
  • Reported EC50 of 0.32 μM in Ca2+ mobilization assay.
  • High purity suitable for research applications.
  • Stable powder with defined storage recommendations.
Catalog No. 50-002-62638 Supplier Medchemexpress LLC Supplier No. HY1330115MG
nAChR agonist 1 is a research compound that functions as a potent positive allosteric modulator of the α7 nicotinic acetylcholine receptor. It is brain-permeable and orally efficacious in preclinical assays, with reported activity in FLIPR Ca2+ mobilization studies. Supplied as an off-white to yellow solid for use in in vitro and in vivo research.
  • Potent positive allosteric modulator of α7 nicotinic acetylcholine receptor.
  • Brain-permeable and orally efficacious in preclinical models.
  • Reported EC50 of 0.32 μM in Ca2+ mobilization assay.
  • High purity suitable for research applications.
  • Stable powder with defined storage recommendations.