Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(5-(4-Chlorophenyl)thieno[2,3-d]pyrimidin-4-ylamino)phenol hydrochloride

Catalog No. OTV000517 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000517 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000517 Supplier Sigma Aldrich Supplier No. OTV000517250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Starting material for the preparation of 1-alkyl-2-methylethenes; Michael additions to enones; Synthesis of α,α-unsaturated nitriles

Specifications

Molecular Formula C18H13Cl2N3OS
Linear Formula C18H13Cl2N3OS
Quantity 50 mL
MDL Number MFCD03622291
Molecular Weight (g/mol) 390.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.