Learn More
Medchemexpress LLC 4-[(5,5,8,8-tetramethyl-6,7-dihydronaphthalene-2-carbonyl)amino]benzoic acid | 102121-60-8 | MFCD00673916 | 98.4% | 351.44 | C22H25NO3 | 100 MG
SDP
Catalog No. 5000241707
Encompass_Preferred
AM580 is a potent, selective retinoic acid receptor α (RARα) agonist for research applications. It has reported IC50 of 8 nM and EC50 of 0.36 nM, chemical formula C22H25NO3, molecular weight 351.44, and is supplied as a white to off-white solid; consult the supplier safety data sheet for handling and storage instructions.
  • Potent RARα activation with low nanomolar potency.
  • Suitable for cellular and biochemical assays.
  • High purity for reproducible results.
  • Stable as a powder under recommended storage conditions.
  • Available in multiple small-scale sizes for flexible experiment planning.
Catalog No. 50-002-41707 Supplier Medchemexpress LLC Supplier No. HY10475100MG
AM580 is a potent, selective retinoic acid receptor α (RARα) agonist for research applications. It has reported IC50 of 8 nM and EC50 of 0.36 nM, chemical formula C22H25NO3, molecular weight 351.44, and is supplied as a white to off-white solid; consult the supplier safety data sheet for handling and storage instructions.
  • Potent RARα activation with low nanomolar potency.
  • Suitable for cellular and biochemical assays.
  • High purity for reproducible results.
  • Stable as a powder under recommended storage conditions.
  • Available in multiple small-scale sizes for flexible experiment planning.