Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5,6,7-Tetrachlorofluorescein, Thermo Scientific™

Catalog No. AAB2563503
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2563503 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2563503 Supplier Thermo Scientific Chemicals Supplier No. B2563503

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6262-21-1
Molecular Formula C20H8Cl4O5
Quantity 1g
MDL Number MFCD00010030
Synonym 4,5,6,7-tetrachlorofluorescein, tetrachlorofluorescein, unii-o73wzt10by, o73wzt10by, 4,5,6,7-tetrachloro-3',6'-dihydroxy-3h-spiro 2-benzofuran-1,9'-xanthen-3-one, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 4,5,6,7-tetrachloro-3',6'-dihydroxy, 4,5,6,7-tetrachloro-3',6'-dihydroxyspiro isobenzofuran-1 3h ,9'-9h xanthene-3-one, fluorescein, tetrachloro, 4,6,7-tetrachloro-fluorescein
InChI Key QCPFFGGFHNZBEP-UHFFFAOYSA-N
SMILES OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1
IUPAC Name 4,5,6,7-tetrachloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol) 470.08
PubChem CID 80424
Formula Weight 470.1
Chemical Name or Material 4,5,6,7-Tetrachlorofluorescein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.