Learn More
Medchemexpress LLC 4,5,6,7-Tetrachloroindan-1,3-dione | 30675-13-9 | 99.8% | 283.92 | 5 MG
SDP
Catalog No. 5000388182
Encompass_Preferred
TCID (4,5,6,7-Tetrachloroindan-1,3-dione) is a potent and selective inhibitor of neuronal ubiquitin C-terminal hydrolase (UCH-L3) with an IC50 of 0.6 μM. It also inhibits UCH-L1 with an IC50 of 75 μM. This compound diminishes glycine transporter GlyT2 ubiquitination in primary neurons from the brainstem and spinal cord.
  • Potent and selective inhibition of UCH-L3.
  • Reduces GlyT2 ubiquitination in neuronal cells.
  • High purity of 99.78% ensures reliability.
  • Useful for studying UCH-L3 activity and GlyT2 ubiquitination in neuronal systems.
Catalog No. 50-003-88182 Supplier Medchemexpress LLC Supplier No. HY186385MG
TCID (4,5,6,7-Tetrachloroindan-1,3-dione) is a potent and selective inhibitor of neuronal ubiquitin C-terminal hydrolase (UCH-L3) with an IC50 of 0.6 μM. It also inhibits UCH-L1 with an IC50 of 75 μM. This compound diminishes glycine transporter GlyT2 ubiquitination in primary neurons from the brainstem and spinal cord.
  • Potent and selective inhibition of UCH-L3.
  • Reduces GlyT2 ubiquitination in neuronal cells.
  • High purity of 99.78% ensures reliability.
  • Useful for studying UCH-L3 activity and GlyT2 ubiquitination in neuronal systems.