Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,5,6,7-tetrachloroindan-1,3-dione | 30675-13-9 | MFCD00239209 | 283.92 g/mol | C9H2Cl4O2 | 10 MG
SDP

Catalog No. 5000297928
Encompass_Preferred

TCID (4,5,6,7-tetrachloroindan-1,3-dione) is a small-molecule research inhibitor of ubiquitin C-terminal hydrolase L3 (UCH-L3), used to probe deubiquitinase activity and cell cycle/DNA damage pathways. It is supplied in milligram-scale quantities for biochemical and cellular studies and includes documented solubility and storage recommendations for research use.

  • CAS number: 30675-13-9
  • Molecular formula: C9H2Cl4O2
  • Molecular weight: 283.92 g/mol
  • Target: deubiquitinase UCH-L3
  • Pack sizes: 1 mg, 5 mg, 10 mg
  • In vitro solubility: DMSO 20 mg/mL (may require ultrasonic)
  • Storage: powder -20°C (3 years) or 4°C (2 years)

Catalog No. 50-002-97928 Supplier Medchemexpress LLC Supplier No. HY1863810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCID (4,5,6,7-tetrachloroindan-1,3-dione) is a small-molecule research inhibitor of ubiquitin C-terminal hydrolase L3 (UCH-L3), used to probe deubiquitinase activity and cell cycle/DNA damage pathways. It is supplied in milligram-scale quantities for biochemical and cellular studies and includes documented solubility and storage recommendations for research use.

  • CAS number: 30675-13-9
  • Molecular formula: C9H2Cl4O2
  • Molecular weight: 283.92 g/mol
  • Target: deubiquitinase UCH-L3
  • Pack sizes: 1 mg, 5 mg, 10 mg
  • In vitro solubility: DMSO 20 mg/mL (may require ultrasonic)
  • Storage: powder -20°C (3 years) or 4°C (2 years)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.