Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,5,6,7-Tetrafluoro-1-hydroxy-1,2-benziodoxole-3(1H)-one | 954373-94-5 | 95.9% | 335.98 | 250 MG
SDP

Catalog No. 5000402241
Encompass_Preferred

4,5,6,7-Tetrafluoro-1-hydroxy-1,2-benziodoxole-3(1H)-one is a drug intermediate for the synthesis of various active compounds.

  • Molecular formula C7HF4IO3
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-02241 Supplier Medchemexpress LLC Supplier No. 250MGHY22018
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4,5,6,7-Tetrafluoro-1-hydroxy-1,2-benziodoxole-3(1H)-one is a drug intermediate for the synthesis of various active compounds.

  • Molecular formula C7HF4IO3
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.